2-(Difluormethoxy)phenylacetonitril

Lieferant: FLUOROCHEM

007870-1G 007870-5G
FLUO007870-1GEA 30.8 EUR
FLUO007870-1G FLUO007870-5G
2-(Difluormethoxy)phenylacetonitril
2-(Difluormethoxy)phenylacetonitril
Formel: C₉H₇F₂NO
MDL: MFCD00236240
CAS-Nummer: 41429-22-5
UN: 3276
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR