3-(Difluormethoxy)phenylacetonitril

Lieferant: FLUOROCHEM

Synonyme: 2-(3-(Difluoromethoxy)phenyl)acetonitrile

007871-1G 007871-2.5G
FLUO007871-1GEA 488 EUR
FLUO007871-1G FLUO007871-2.5G
3-(Difluormethoxy)phenylacetonitril
3-(Difluormethoxy)phenylacetonitril
Formel: C₉H₇F₂NO
Molecular Weight: 183,15 g/mol
MDL: MFCD00236241
CAS-Nummer: 41429-18-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR