Digitonin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

SIALD5628-5GEA 1800 EUR
SIALD5628-5G SIALD5628-1G
Digitonin, Sigma-Aldrich®
Digitonin
Formel: C₅₆H₉₂O₂₉
Molecular Weight: 1229,312 g/mol
MDL: MFCD00077729
CAS-Nummer: 11024-24-1
EINECS: 234-255-6
UN: 3462
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR