5'-Chlor-2'-methoxyacetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(5-Chloro-2-methoxyphenyl)ethanone

019292-5G 019292-25G
FLUO019292-5GEA 130 EUR
FLUO019292-5G FLUO019292-25G
5'-Chlor-2'-methoxyacetophenon
5'-Chlor-2'-methoxyacetophenon
Formel: C₉H₉ClO₂
Molecular Weight: 184.6196 g/mol
MDL: MFCD00174339
CAS-Nummer: 6342-64-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR