1,5-Dibrom-2,4-difluorbenzol
Lieferant: COMBI-BLOCKS
OT-1604-5G
OT-1604-25G
COBBOT-1604-5GEA
63
EUR
COBBOT-1604-5G
COBBOT-1604-25G
1,5-Dibrom-2,4-difluorbenzol
1,5-Dibrom-2,4-difluorbenzol
Formel:
C₆H₂Br₂F₂ Molecular Weight: 271,8849 g/mol |
MDL:
MFCD08458122 CAS-Nummer: 28342-75-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...