1,3-Dibrom-5-(trifluormethoxy)benzol

Lieferant: FLUOROCHEM

023874-1G 023874-5G
FLUO023874-1GEA 31.1 EUR
FLUO023874-1G FLUO023874-5G
1,3-Dibrom-5-(trifluormethoxy)benzol
1,3-Dibromo-5-(trifluoromethoxy)benzene
Formel: C₇H₃Br₂F₃O
MDL: MFCD00075280
CAS-Nummer: 207226-31-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR