2-Brombenzylamin Hydrochlorid 97%
Lieferant: Thermo Scientific
Achtung
A18151.06 A18151.14
A18151.06EA 57.6 EUR
A18151.06 A18151.14
2-Brombenzylamin Hydrochlorid 97%
2-Brombenzylamin Hydrochlorid
Purity: 97% CAS No.: 5465-63-4 Molecular Formula: C7H8BrN.HCl Formula Weight: 222.52 Melting Point: 227-230° Beilstein Reference: 3913456
Formel: C₇H₉BrClN
Molecular Weight: 222,52 g/mol
Schmelzpunkt: 227…230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012853
CAS-Nummer: 5465-63-4