3-Brombenzylamin Hydrochlorid 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(3-Bromophenyl)methanamine hydrochloride
A16132.06
A16132.14
A16132.06EA
33
EUR
A16132.06
A16132.14
3-Brombenzylamin Hydrochlorid 98%
3-Brombenzylamin Hydrochlorid
Purity: 98%
CAS No.: 39959-54-1
Molecular Formula: C7H8BrN.HCl
Formula Weight: 222.52
Melting Point: 218-221°
Beilstein Reference: 3913185
Formel:
C₇H₉BrClN Molecular Weight: 222,52 g/mol Schmelzpunkt: 218…221 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012856 CAS-Nummer: 39959-54-1 |