2-Fluor-N-methylbenzylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(2-Fluorophenyl)-N-methylmethanamine, (2-Fluorobenzyl)methylamine

M-Clarity™ Qualitätsstufe = MQ100

631094-1G 631094-5G
SIAL631094-1GEA 53.9 EUR
SIAL631094-1G SIAL631094-5G
2-Fluor-N-methylbenzylamin, Sigma-Aldrich®
2-Fluor-N-methylbenzylamin
Formel: C₈H₁₀FN
Molecular Weight: 139,17 g/mol
MDL: MFCD04629633
CAS-Nummer: 399-30-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR