3-Fluor-N-methylbenzylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (3-Fluorobenzyl)methylamine, 1-(3-Fluorophenyl)-N-methylmethanamine

M-Clarity™ Qualitätsstufe = MQ100

631108-1G 631108-5G
SIAL631108-1GEA 26 EUR
SIAL631108-1G SIAL631108-5G
3-Fluor-N-methylbenzylamin, Sigma-Aldrich®
3-Fluor-N-methylbenzylamin
Formel: C₈H₁₀FN
Molecular Weight: 139,17 g/mol
MDL: MFCD04623547
CAS-Nummer: 90389-84-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR