4-Fluor-N-methylbenzylamin 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
N-Methyl-4-fluorobenzylamine
H54369.03
H54369.06
H54369.03EA
35.3
EUR
H54369.03
H54369.06
4-Fluor-N-methylbenzylamin 97%
4-Fluor-N-methylbenzylamin
Purity: 97%
CAS No.: 405-66-3
Molecular Formula: C8H10FN
Formula Weight: 139.17
Flash Point: 71°(160°F)
Refractive Index: 1.499
Boiling Point: 180-181°
MDL No.: MFCD03839848
Formel:
C₈H₁₀FN Molecular Weight: 139,17 g/mol Siedepunkt: 180…181 °C Dichte: 1,056 Flammpunkt: 71 °C (160 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03839848 CAS-Nummer: 405-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...