4-Fluor-N-methylbenzylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Methyl-4-fluorobenzylamine

593842-1G 593842-5G
SIAL593842-1GEA 45.4 EUR
SIAL593842-1G SIAL593842-5G
4-Fluor-N-methylbenzylamin, Sigma-Aldrich®
4-Fluor-N-methylbenzylamin
Formel: C₈H₁₀FN
Molecular Weight: 139,17 g/mol
MDL: MFCD03839848
CAS-Nummer: 405-66-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR