1,2-Diiod-4-fluorbenzol

Lieferant: CHEMPUR

FL13307.POR 013307-25G 013307-5G
CHMPFL13307.POREA 0 EUR
CHMPFL13307.POR FLUO013307-25G FLUO013307-5G
1,2-Diiod-4-fluorbenzol
1,2-Diiodo-4-fluorobenzene
Formel: C₆H₃FI₂
MDL: MFCD00042576
CAS-Nummer: 203059-85-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR