3-(N,N-Dimethylamino)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-(N,N-Dimethylamino)phenylboronic acid, pinacol ester, N,N-Dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-(Dimethylamino)phenylboronic acid pinacol ester

PN-8674-5G PN-8674-1G
COBBPN-8674-5GEA 1490 EUR
COBBPN-8674-5G COBBPN-8674-1G
3-(N,N-Dimethylamino)phenylboronsäurepinakolester
3-(N,N-Dimethylamino)phenylboronsäurepinakolester
Formel: C₁₄H₂₂BNO₂
Molecular Weight: 247,1400 g/mol
MDL: MFCD06795669
CAS-Nummer: 325142-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR