5-Chlor-2-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5287-5G PN-5287-1G
COBBPN-5287-5GEA 1490 EUR
COBBPN-5287-5G COBBPN-5287-1G
5-Chlor-2-nitrophenylboronsäurepinakolester
5-Chlor-2-nitrophenylboronsäurepinakolester
Formel: C₁₂H₁₅BClNO₄
MDL: MFCD11053894
CAS-Nummer: 1073353-99-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR