2-(Trifluormethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4,4,5,5-Tetramethyl-2-(2-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane

PN-2625-5G PN-2625-1G
COBBPN-2625-5GEA 0 EUR
COBBPN-2625-5G COBBPN-2625-1G
2-(Trifluormethyl)phenylboronsäurepinakolester
2-(Trifluormethyl)phenylboronsäurepinakolester
Formel: C₁₃H₁₆BF₃O₂
Molecular Weight: 272.0712 g/mol
MDL: MFCD06795676
CAS-Nummer: 1073339-21-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR