3-(Trifluormethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4,4,5,5-Tetramethyl-2-[3-(trifluoromethyl)phenyl]-1,3,2-dioxaborolane, 3-(Trifluoromethyl)phenylboronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(3-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)benzene

PN-2378-5G PN-2378-1G
COBBPN-2378-5GEA 166 EUR
COBBPN-2378-5G COBBPN-2378-1G
3-(Trifluormethyl)phenylboronsäurepinakolester
3-(Trifluormethyl)phenylboronsäurepinakolester
Formel: C₁₃H₁₆BF₃O₂
Molecular Weight: 272.0712 g/mol
MDL: MFCD05863925
CAS-Nummer: 325142-82-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR