3-Fluor-4-(trifluormethyl)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-(3-Fluoro-4-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester
PN-8824-5G
PN-8824-1G
COBBPN-8824-5GEA
240
EUR
COBBPN-8824-5G
COBBPN-8824-1G
3-Fluor-4-(trifluormethyl)phenylboronsäurepinakolester
3-Fluor-4-(trifluormethyl)phenylboronsäurepinakolester
Formel:
C₁₃H₁₅BF₄O₂ Molecular Weight: 290,06 g/mol |
MDL:
MFCD09878544 CAS-Nummer: 445303-67-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...