3-Fluor-4-methylphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
3-Fluoro-4-methylbenzeneboronic acid pinacol ester, 2-(3-Fluoro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PN-2209-5G
PN-2209-1G
COBBPN-2209-5GEA
451
EUR
COBBPN-2209-5G
COBBPN-2209-1G
3-Fluor-4-methylphenylboronsäurepinakolester
3-Fluor-4-methylphenylboronsäurepinakolester
Formel:
C₁₃H₁₈BFO₂ Molecular Weight: 236.09 g/mol |
MDL:
MFCD09972178 CAS-Nummer: 903895-56-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...