4-(Trifluormethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4,4,5,5-Tetramethyl-2-(4-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane, 4-Trifluoromethylphenylboronic acid pinacol ester

PN-2673-5G PN-2673-1G
COBBPN-2673-5GEA 181 EUR
COBBPN-2673-5G COBBPN-2673-1G
4-(Trifluormethyl)phenylboronsäurepinakolester
4-(Trifluormethyl)phenylboronsäurepinakolester
Formel: C₁₃H₁₆BF₃O₂
Molecular Weight: 272.0712 g/mol
MDL: MFCD05863924
CAS-Nummer: 214360-65-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR