4-Fluor-2-methylphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(4-Fluoro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Fluoro-2-methylbenzeneboronic acid pinacol ester

PN-2204-25G PN-2204-1G PN-2204-5G
COBBPN-2204-25GEA 247 EUR
COBBPN-2204-25G COBBPN-2204-1G COBBPN-2204-5G
4-Fluor-2-methylphenylboronsäurepinakolester
4-Fluor-2-methylphenylboronsäurepinakolester
Formel: C₁₃H₁₈BFO₂
Molecular Weight: 236.09 g/mol
MDL: MFCD05863911
CAS-Nummer: 815631-56-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR