4-Chlor-3-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chloro-3-nitrobenzeneboronic acid

BB-2037-1G BB-2037-5G
COBBBB-2037-1GEA 56.8 EUR
COBBBB-2037-1G COBBBB-2037-5G
4-Chlor-3-nitrophenylboronsäure
4-Chlor-3-nitrophenylboronsäure
Formel: C₆H₅BClNO₄
Molecular Weight: 201,37 g/mol
MDL: MFCD02258950
CAS-Nummer: 151169-67-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR