4-Isobutylphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Isobutylphenylboronic acid, pinacol ester, 2-(4-Isobutylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-2441-5G PN-2441-1G
COBBPN-2441-5GEA 375 EUR
COBBPN-2441-5G COBBPN-2441-1G
4-Isobutylphenylboronsäurepinakolester
4-Isobutylphenylboronsäurepinakolester
Formel: C₁₆H₂₅BO₂
Molecular Weight: 260,1795 g/mol
MDL: MFCD05663842
CAS-Nummer: 1033753-01-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR