3,5-Di-tert-butylbenzylbromid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-(Bromomethyl)-3,5-di-tert-butylbenzene
M-Clarity™ Qualitätsstufe = MQ100
571474-1G
571474-5G
SIAL571474-1GEA
80.5
EUR
SIAL571474-1G
SIAL571474-5G
3,5-Di-tert-butylbenzylbromid, Sigma-Aldrich®
3,5-Di-tert-butylbenzylbromid
Formel:
C₁₅H₂₃Br Molecular Weight: 283,2471 g/mol |
MDL:
MFCD03701623 CAS-Nummer: 62938-08-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...