1,3,5-Tris(brommethyl)benzol, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
657336-10G
657336-5G
SIAL657336-10GEA
248
EUR
SIAL657336-10G
SIAL657336-5G
1,3,5-Tris(brommethyl)benzol, Sigma-Aldrich®
1,3,5-Tris(brommethyl)benzol
Formel:
C₉H₉Br₃ Molecular Weight: 356,88 g/mol |
MDL:
MFCD00182500 CAS-Nummer: 18226-42-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...