Lithiumhexafluoroantimonat
Lieferant: FLUOROCHEM
|
Achtung
|
002846-25G
002846-100G
FLUO002846-25GEA
467
EUR
FLUO002846-25G
FLUO002846-100G
Lithiumhexafluoroantimonat
Lithium hexafluoroantimonate
Formel:
F₆LiSb Molecular Weight: 242.68 g/mol |
MDL:
MFCD00042319 CAS-Nummer: 18424-17-4 UN: 1549 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...