Lanthan(III)carbonat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)

Lieferant: Thermo Scientific
Achtung

16639.14 16639.22 16639.36
16639.14EA 22.9 EUR
16639.14 16639.22 16639.36
Lanthan(III)carbonat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lanthan(III)carbonat Hydrat

Purity: 99.9% CAS No.: 54451-24-0 Molecular Formula: La2(CO3)3.xH2O Formula Weight: 457.85(anhy) Physical Form: Powder Density: 2.6-2.7 Merck Reference: 14,5364 MDL No.: MFCD00011069

Formel: La₂(CO₃)₃.xH₂O
Molecular Weight: 457,85 g/mol (anhy)
Dichte: 2,6…2.7
Lagertemperatur: Raumtemperatur
MDL: MFCD00011069
CAS-Nummer: 54451-24-0
EINECS: 209-599-5
Merck Index: 14,5364

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR