Mangan(II)sulfid ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
12835.06
12835.14
12835.22
12835.06EA
43.3
EUR
12835.06
12835.14
12835.22
Mangan(II)sulfid ≥99,9% (Metall-Basis)
Manganese(II) sulphide
Purity: 99.9%
CAS No.: 18820-29-6
Molecular Formula: MnS
Formula Weight: 87.00
Physical Form: -325 Mesh Powder
Density: 3.99
Appearance: Green to pink
Melting Point: 1610° dec.
Merck Reference: 14,5740
MDL No.: MFCD00014210
Formel:
MnS Molecular Weight: 87.00 g/mol Schmelzpunkt: 1610 °C (Zers.) Dichte: 3.99 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014210 CAS-Nummer: 18820-29-6 EINECS: 242-599-3 Merck Index: 14,5740 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...