4-Isopropoxy-2-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Isopropoxy-2-methylbenzeneboronic acid

BB-2794-1G BB-2794-5G
COBBBB-2794-1GEA 158 EUR
COBBBB-2794-1G COBBBB-2794-5G
4-Isopropoxy-2-methylphenylboronsäure
4-Isopropoxy-2-methylphenylboronsäure
Formel: C₁₀H₁₅BO₃
Molecular Weight: 194.04 g/mol
MDL: MFCD07363788
CAS-Nummer: 871126-21-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR