4-Isopropoxy-3-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Isopropoxy-3-methylbenzeneboronic acid

BB-2741-5G BB-2741-25G BB-2741-1G
COBBBB-2741-5GEA 304 EUR
COBBBB-2741-5G COBBBB-2741-25G COBBBB-2741-1G
4-Isopropoxy-3-methylphenylboronsäure
4-Isopropoxy-3-methylphenylboronsäure
Formel: C₁₀H₁₅BO₃
Molecular Weight: 194.04 g/mol
MDL: MFCD06659871
CAS-Nummer: 850568-09-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR