3-Brom-2-fluor-5-methylphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Bromo-2-fluoro-5-methylphenyl)boronic acid
BB-5070-5G
BB-5070-1G
COBBBB-5070-5GEA
303
EUR
COBBBB-5070-5G
COBBBB-5070-1G
3-Brom-2-fluor-5-methylphenylboronsäure
3-Brom-2-fluor-5-methylphenylboronsäure
Formel:
C₇H₇BBrFO₂ Molecular Weight: 232.8400 g/mol |
MDL:
MFCD09878334 CAS-Nummer: 957066-00-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...