3-Brom-5-(trifluormethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Bromo-5-(trifluoromethyl)benzeneboronic acid
BB-3460-5G
BB-3460-1G
COBBBB-3460-5GEA
399
EUR
COBBBB-3460-5G
COBBBB-3460-1G
3-Brom-5-(trifluormethyl)phenylboronsäure
3-Brom-5-(trifluormethyl)phenylboronsäure
Formel:
C₇H₅BBrF₃O₂ Molecular Weight: 268.82 g/mol |
MDL:
MFCD08436051 CAS-Nummer: 913835-64-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...