Piperidin 0.3 mol/l in Methanol
Lieferant: Bernd Kraft
Synonyme:
Hexahydropyridin, Hexazan, Pentamethylenimin
05479.4000
05479.2000
05479.3000
KRAF05479.4000EA
0
EUR
KRAF05479.4000
KRAF05479.2000
KRAF05479.3000
Piperidin 0.3 mol/l in Methanol
Piperidin
Formel:
C₅H₁₁N Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005979 CAS-Nummer: 110-89-4 EINECS: 203-813-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...