2-Nitro-5-(1-piperidinyl)phenol

Lieferant: CHEMPUR

BTB05906.1 BTB05906.10 BTB05906.50
CHMPBTB05906.1EA 0 EUR
CHMPBTB05906.1 CHMPBTB05906.10 CHMPBTB05906.50
2-Nitro-5-(1-piperidinyl)phenol
2-Nitro-5-(1-piperidinyl)phenol
Formel: C₁₁H₁₄N₂O₃
Molecular Weight: 224.24 g/mol
MDL: MFCD00052663
CAS-Nummer: 157831-75-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR