2,6-Difluor-DL-phenylalanin

Lieferant: FLUOROCHEM

007874-1G 007874-2.5G
FLUO007874-1GEA 249 EUR
FLUO007874-1G FLUO007874-2.5G
2,6-Difluor-DL-phenylalanin
2,6-Difluor-DL-phenylalanin
Formel: C₉H₉F₂NO₂
MDL: MFCD00061304
CAS-Nummer: 32133-39-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR