6,8-Dichlor-3-cyanchromon
Lieferant: CHEMPUR
Synonyme:
6,8-Dichloro-3-cyanochromone, 6,8-Dichloro-4-oxo-4H-1-benzopyran-3-carbonitrile, 6,8-Dichlorochromone-3-carbonitrile
FL12019.POR
012019-5G
CHMPFL12019.POREA
0
EUR
CHMPFL12019.POR
FLUO012019-5G
6,8-Dichlor-3-cyanchromon
6,8-Dichlor-3-cyanchromon
Formel:
C₁₀H₃Cl₂NO₂ |
MDL:
MFCD00192004 CAS-Nummer: 72798-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...