D(-)-Glutamin ≥99%

Lieferant: Thermo Scientific

Synonyme: D(-)-2-Amino-1,5-pentandisäure-5-amidD(-)-Glutaminsäure-5-amid(R)-(-)-Glutamin(R)-(-)-2-Amino-1,5-pentandisäure-5-amid(R)-(-)-Glutaminsäure-5-amidH-D-Gln-OH

J60784.03EA 42.7 EUR
J60784.03 J60784.14 J60784.06
D(-)-Glutamin ≥99%
D(-)-Glutamin

Kinase inhibitor; has antivasospastic properties

Formel: C₅H₁₀N₂O₃
Molecular Weight: 146,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00065607
CAS-Nummer: 5959-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR