3-Methoxyanthranilsäure

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Amino-3-methoxybenzoesäure

J64244.06 J64244.14
J64244.06EA 129 EUR
J64244.06 J64244.14
3-Methoxyanthranilsäure
3-Methoxyanthranilsäure
Formel: C₈H₉NO₃
Molecular Weight: 167.16 g/mol
Schmelzpunkt: 169…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075178
CAS-Nummer: 3177-80-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR