2-Amino-4-chlorpyridin 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Chlorpyridin-2-amin, 4-Chlor-2-pyridylamin

H59130.06 H59130.14
H59130.06EA 61.8 EUR
H59130.06 H59130.14
2-Amino-4-chlorpyridin 97%
2-Amino-4-chlorpyridin
Formel: C₅H₅ClN₂
Molecular Weight: 128.56 g/mol
Schmelzpunkt: 129…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04113820
CAS-Nummer: 19798-80-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR