Bismut(III)oxid ≥99,9%, Nano-Pulver

Lieferant: Thermo Fisher Scientific
Achtung

46314.22 46314.36 46314.14
46314.22EA 101 EUR
46314.22 46314.36 46314.14
Bismut(III)oxid ≥99,9%, Nano-Pulver
Bismut(III)oxid

Purity: 99.9% CAS No.: 1304-76-3 Molecular Formula: Bi2O3 Formula Weight: 465.96 Physical Form: 80-200nm APS Powder, S.A. 3.8-5.5m2/g Density: 8.9 Appearance: Yellow Melting Point: 817° Boiling Point: ca 1890° Merck Reference: 14,1273 MDL No.: MFCD00003462

Formel: BiO₃
Siedepunkt: ∼1890 °C
Schmelzpunkt: 817 °C
Dichte: 8,9
Lagertemperatur: Raumtemperatur
MDL: MFCD00003462
CAS-Nummer: 1304-76-3
EINECS: 215-134-7
Merck Index: 14,1273

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR