Tetraethylammoniumchlorid

Lieferant: Thermo Scientific
Achtung

Synonyme: Tetrylammoniumchlorid

J62507.14 J62507.22
J62507.14EA 36.8 EUR
J62507.14 J62507.22
Tetraethylammoniumchlorid
Tetraethylammoniumchlorid

CAS No.: 56-34-8 Molecular Formula: C8H20ClN Formula Weight: 165.71 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White to pale yellow Solubility: Soluble in water, ethanol, chloroform, or acetone Beilstein Reference: 3563247 Merck Reference: 14,9200 MDL No.: MFCD00011828


Phase transfer catalyst

Formel: C₈H₂₀ClN
Molecular Weight: 165,71 g/mol
Schmelzpunkt: 285 °C
Dichte: 1,08 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011828
CAS-Nummer: 56-34-8
EINECS: 200-267-5
Merck Index: 14,9200

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR