N-Lauroylsarkosin Natriumsalz 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
N-Dodecanoyl-N-methylglycin Natriumsalz, Lauroylsarkosinat, Natrium-N-lauroylsarkosinat, Sarkosyl
J60040.09
J60040.18
J60040.09EA
29.5
EUR
J60040.09
J60040.18
N-Lauroylsarkosin Natriumsalz 95%
N-Lauroylsarkosin Natriumsalz
Purity: 95%
CAS No.: 137-16-6
Molecular Formula: C15H29NNaO3
Formula Weight: 293.39
Physical Form: Crystalline powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 44-47°
Beilstein Reference: 5322974
Merck Reference: 14,4368
MDL No.: MFCD00042728
Used in the cell lysis step of RNA purification. Anionic detergent
Formel:
H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa Molecular Weight: 293,38 g/mol Schmelzpunkt: 44…47 °C Flammpunkt: >150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042728 CAS-Nummer: 137-16-6 EINECS: 205-281-5 UN: 2811 ADR: 6.1,II Merck Index: 14,4368 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...