N,N-Dimethylformamiddimethylacetal

Lieferant: COMBI-BLOCKS
Gefahr

Synonyme: 1,1-Dimethoxytrimethylamin

OT-0501-25G OT-0501-100G
COBBOT-0501-25GEA 50.9 EUR
COBBOT-0501-25G COBBOT-0501-100G
N,N-Dimethylformamiddimethylacetal
N,N-Dimethylformamiddimethylacetal
Formel: C₅H₁₃NO₂
Molecular Weight: 119,16 g/mol
Siedepunkt: 102 °C (1013 hPa)
Dichte: 0,893 g/cm³ (20 °C)
Flammpunkt: 6 °C
MDL: MFCD00008482
CAS-Nummer: 4637-24-5
EINECS: 225-063-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR