Methylenblau Trihydrat
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3,7-Bis(dimethylamino)phenothiaziniumchlorid Trihydrat, N,N,N′,N′-Tetramethylthioninchlorid Trihydrat, Methylthioniniumchlorid Trihydrat, C.I.52015
J60823.14
J60823.22
J60823.30
J60823.14EA
38.2
EUR
J60823.14
J60823.22
J60823.30
Methylenblau Trihydrat
Methylenblau Trihydrat
CAS No.: 7220-79-3
Molecular Formula: C16H18ClN3S.3H2O
Formula Weight: 373.90 (319.86anhy)
Physical Form: Crystalline Powder
Appearance: Green
Odour: Odourless
Beilstein Reference: 3599847
Merck Reference: 14,6060
MDL No.: MFCD00150008
Sensitive stain for RNA and ribonuclease
Formel:
C₁₆H₁₈ClN₃S·3H₂O Molecular Weight: 373,9 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150008 CAS-Nummer: 7220-79-3 EINECS: 200-515-2 Merck Index: 14,6060 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...