o-Phenylendiamin Dihydrochlorid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98+%
CAS No.: 615-28-1
Molecular Formula: C6H8N2.2HCl
Formula Weight: 181.07
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Beilstein Reference: 3912045
UN No.: 2811
MDL No.: MFCD00012966
A peroxidase substrate for ELISA procedures
Formel:
C₆H₁₀Cl₂N₂ Schmelzpunkt: 205 °C |
MDL:
MFCD00012966 CAS-Nummer: 615-28-1 EINECS: 210-418-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...