Neutralrot ACS
Lieferant: Thermo Fisher Scientific
Synonyme:
Basisrot 5, 2-Amino-8-(dimethylamino)-3-methylphenaziniumchlorid, N8,N8,3-Trimethyl-2,8-phenazindiamin Monohydrochlorid
J62643.18
J62643.30
J62643.18EA
38.7
EUR
J62643.18
J62643.30
Neutralrot ACS
Neutralrot
CAS No.: 553-24-2
Molecular Formula: C15H17ClN4
Formula Weight: 288.78
Physical Form: Powder
Appearance: Dark green to black
Melting Point: 290° dec.
Beilstein Reference: 3918740
Merck Reference: 14,6488
MDL No.: MFCD00012651
Nuclear counterstain useful in pH range 6.8 - 8.0
Formel:
C₁₅H₁₇ClN₄ Molecular Weight: 288.78 g/mol Schmelzpunkt: 290 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012651 CAS-Nummer: 553-24-2 EINECS: 209-035-8 Merck Index: 14,6488 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...