Neutralrot
Lieferant: Thermo Fisher Scientific
Synonyme: Basisrot 5, 2-Amino-8-(dimethylamino)-3-methylphenaziniumchlorid, N8,N8,3-Trimethyl-2,8-phenazindiamin Monohydrochlorid
J63054.03 J63054.06 J63054.14
J63054.03EA 35.4 EUR
J63054.03 J63054.06 J63054.14
Neutralrot
Neutralrot
CAS No.: 553-24-2 Molecular Formula: C15H17ClN4 Formula Weight: 288.78 Physical Form: Powder Appearance: Dark green to black Melting Point: 290° dec. Beilstein Reference: 3918740 Merck Reference: 14,6488 MDL No.: MFCD00012651
Nuclear counterstain useful in pH range 6.8 - 8.0
Formel: C₁₅H₁₇ClN₄
Molecular Weight: 288.78 g/mol
Schmelzpunkt: 290 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012651
CAS-Nummer: 553-24-2
EINECS: 209-035-8
Merck Index: 12,06571