L-α-Phenylalanin Reagenz für die Zellkultur
Lieferant: Thermo Scientific
Synonyme: H-Phe-OH, H-L-Phe-OH, L-α-Phenylalanin, 3-Phenyl-L(-)-alanin, 3-Phenyl-L-alanin, (S)-(-)-2-Amino-3-phenylpropansäure
J63925.22 J63925.30 J63925.36
J63925.22EA 99.4 EUR
J63925.22 J63925.30 J63925.36
L-α-Phenylalanin Reagenz für die Zellkultur
L-α-Phenylalanin
CAS No.: 63-91-2 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Physical Form: Powder Appearance: White to off-white Solubility: Soluble in water. Aqueous solubility increases with warming. Also soluble in methanol, ethanol or 1M NH4OH Beilstein Reference: 1910408 Merck Reference: 14,7271 MDL No.: MFCD00064227
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
Siedepunkt: 313 °C (1013 hPa)
Schmelzpunkt: 173 °C
Dichte: 0,754 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064227
CAS-Nummer: 63-91-2
EINECS: 200-568-1
Merck Index: 14,7271