Rhodamin 6G

Lieferant: Thermo Scientific
Achtung

Synonyme: 9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthyliumchlorid

J62315.14 J62315.22
J62315.14EA 42.1 EUR
J62315.14 J62315.22
Rhodamin 6G
Rhodamin 6G

CAS No.: 989-38-8 Molecular Formula: C28H31ClN2O3 Formula Weight: 479.02 Physical Form: Powder Appearance: Red to brown Beilstein Reference: 3900071


Suitable for fluorescent labelling

Formel: C₂₈H₃₁ClN₂O₃
Molecular Weight: 479,02 g/mol
Siedepunkt: ∼200 °C (1013 hPa)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012665
CAS-Nummer: 989-38-8
EINECS: 213-584-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR