Nitroblau-Tetrazoliumchlorid (NBT) ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylen)ditetrazoliumdichlorid, 3,3'-(3,3'-Dimethoxy-4,4'-biphenylen)di(2-(4-nitrophenyl)-5-phenyl-2H-tetrazoliumchlorid), 3,3'-(3,3'-Dimethoxy-4,4'-biphenylen)bis(2-(4-nitrophenyl)-5-phenyl-2H-tetrazoliumchlorid)
Purity: 99%
CAS No.: 298-83-9
Molecular Formula: C40H30Cl2N10O6
Formula Weight: 817.65
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Melting Point: ca 190° dec.
Beilstein Reference: 4115923
MDL No.: MFCD00012159
Intense blue dye used in histochemistry determinations
Formel:
C₄₀H₃₀Cl₂N₁₀O₆ Molecular Weight: 817,65 g/mol Schmelzpunkt: ∼ 190 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00012159 CAS-Nummer: 298-83-9 EINECS: 206-067-4 |